missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R3A Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | PPP1R3A |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18388111
|
Novus Biologicals
NBP3-05174-20UL |
20 μL |
£151.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18350072
|
Novus Biologicals
NBP3-05174-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PPP1R3A Polyclonal specifically detects PPP1R3A in Human, Mouse samples. It is validated for Western Blot.Specifications
| PPP1R3A | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 5506 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| glycogen and sarcoplasmic reticulum binding subunit, skeletal muscle, GM, PP1G, PPP1R3, protein phosphatase 1 glycogen- associated regulatory subunit, Protein phosphatase 1 glycogen-associated regulatory subunit, protein phosphatase 1 regulatory subunit 3A, protein phosphatase 1, regulatory (inhibitor) subunit 3A, protein phosphatase 1, regulatory (inhibitor) subunit 3A (glycogen andsarcoplasmic reticulum binding subunit, skeletal muscle), Protein phosphatase type-1 glycogen targeting subunit, RG1, serine/threonine specific protein phosphatase, type-1 protein phosphatase skeletal muscle glycogen targeting subunit | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 900-1080 of human PPP1R3A (NP_002702.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title