missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPP1R2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566256
This item is not returnable.
View return policy
Description
Immunogen sequence: EPSTPYHSMMG DDEDACSDTE ATEAMAPDIL ARKLAAAEGL EPKYRIQEQE SSGEEDSDLS PEEREKKRQF EMKRKLHYNE GLNIK Highest antigen sequence identity to the following orthologs - mouse 84%, rat 81%.
Protein phosphatase-1 (PP1) is one of the main eukaryotic serine/threonine phosphatases. The protein encoded by this gene binds to the catalytic subunit of PP1, strongly inhibiting its activity. Ten related pseudogenes have been found throughout the human genome. Several splice variants encoding different isoforms have been found for this gene.
Specifications
| PPP1R2 | |
| Polyclonal | |
| Unconjugated | |
| PPP1R2 | |
| 0610025N14Rik; 2310007G06Rik; 4930440J04Rik; 5430408E15Rik; D16Ertd248e; Ipp2; IPP-2; phosphatase inhibitor 2; phosphoprotein phosphatase; PPP1R2; protein phosphatase 1 regulatory inhibitor subunit 2; protein phosphatase 1, regulatory (inhibitor) subunit 2; protein phosphatase 1, regulatory subunit 2; protein phosphatase inhibitor 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5504 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P41236 | |
| PPP1R2 | |
| Recombinant Human PPP1R2. Recombinant protein control fragment (Product #RP-106937). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction