missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPP1R18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560812
This item is not returnable.
View return policy
Description
Immunogen sequence: LSPGESAYQK LGLTEAHKWR PDSRESQEQS LVQLEATEWR LRSGEERQDY SEECGRKEEW PVPGVAPKET AELSETLTRE AQGNSSAGVE Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 57%.
PHTNS interacts with regulatory subunits that target the enzyme to different cellular locations and change its activity toward specific substrates. Phostensin is a regulatory subunit that targets PP1 to F-actin cytoskeleton.
Specifications
| PPP1R18 | |
| Polyclonal | |
| Unconjugated | |
| PPP1R18 | |
| HKMT1098; KIAA1949; Phostensin; PPP1R18; protein phosphatase 1 F-actin cytoskeleton targeting subunit; Protein phosphatase 1 F-actin cytoskeleton-targeting subunit; Protein phosphatase 1 regulatory subunit 18; protein phosphatase 1, regulatory subunit 18 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 170954 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6NYC8 | |
| PPP1R18 | |
| Recombinant protein corresponding to Human PPP1R18. Recombinant protein control fragment (Product #RP-99603). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction