missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R16B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539366
Code : NEW
Additional Details : Weight : 0.00900kg
Description
The protein encoded by this gene is membrane-associated and contains five ankyrin repeats, a protein phosphatase-1-interacting domain, and a carboxy-terminal CAAX box domain. Synthesis of the encoded protein is inhibited by transforming growth factor beta-1. The protein may bind to the membrane through its CAAX box domain and may act as a signaling molecule through interaction with protein phosphatase-1. Alternatively spliced transcript variants encoding different isoforms have been identified in this gene.[provided by RefSeq, Feb 2010].Specifications
PPP1R16B | |
Polyclonal | |
Unconjugated | |
PPP1R16B | |
ANKRD4, KIAA0823 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
228852, 26051 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q96T49, Q8VHQ3 | |
PPP1R16B | |
A synthetic peptide derived from the internal region of human PPP1R16B | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |