missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R15B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14634-1-AP-150UL
This item is not returnable.
View return policy
Description
Maintains low levels of EIF2S1 phosphorylation in unstressed cells by promoting its dephosphorylation by PP1.Specifications
| PPP1R15B | |
| Polyclonal | |
| Unconjugated | |
| PPP1R15B | |
| CREP, PPP1R15B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 108954, 304799, 84919 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5SWA1, Q8BFW3 | |
| Ppp1r15b | |
| PPP1R15B Fusion Protein Ag6213 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction