missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPP1R15B Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551128
This item is not returnable.
View return policy
Description
PPP1R15B Polyclonal Antibody detects endogenous levels of PPP1R15B protein.
Maintains low levels of EIF2S1 phosphorylation in unstressed cells by promoting its dephosphorylation by PP1.
Specifications
| PPP1R15B | |
| Polyclonal | |
| Unconjugated | |
| PPP1R15B | |
| 1810033K10Rik; AI606441; C530022L24Rik; constitutive repressor of eIF2alpha phosphorylation; CReP; MSSGM2; Ppp1r15b; Protein phosphatase 1 regulatory subunit 15B; protein phosphatase 1, regulatory (inhibitor) subunit 15b; protein phosphatase 1, regulatory subunit 15B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84919 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5SWA1 | |
| PPP1R15B | |
| Synthesized peptide derived from the Internal region of human PPP1R15B (aa 411-460). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction