missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R14D/GBPI-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-85484
This item is not returnable.
View return policy
Description
PPP1R14D/GBPI-1 Polyclonal specifically detects PPP1R14D/GBPI-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| PPP1R14D/GBPI-1 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200-1:500 | |
| CPI17-like, FLJ20251, gastrointestinal and brain-specific PP1-inhibitory protein 1, GBPI, GBPI1, GBPI-1, gut and brain phosphatase inhibitor 1, MGC119014, MGC119016, PKC-dependent PP1 inhibitory protein, protein phosphatase 1 regulatory subunit 14D, protein phosphatase 1, regulatory (inhibitor) subunit 14D | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 54866 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| PPP1R14D | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ENPCKKVHWASGRRRTSSTDSESKSHPDSSKIPRSRRPSRLTVKYDRGQLQRWLEMEQWVDAQVQELFQDQATPSEPEIDLEALMDLSTEEQKTQLEAILGNCP | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion