missing translation for 'onlineSavingsMsg'
Learn More

PPP1R14D/GBPI-1 Antibody [DyLight 680], Novus Biologicals™

Product Code. 30761199 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30761199 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30761199 Supplier Novus Biologicals Supplier No. NBP346919FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PPP1R14D/GBPI-1 Polyclonal antibody specifically detects PPP1R14D/GBPI-1 in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R14D/GBPI-1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias CPI17-like, FLJ20251, gastrointestinal and brain-specific PP1-inhibitory protein 1, GBPI, GBPI1, GBPI-1, gut and brain phosphatase inhibitor 1, MGC119014, MGC119016, PKC-dependent PP1 inhibitory protein, protein phosphatase 1 regulatory subunit 14D, protein phosphatase 1, regulatory (inhibitor) subunit 14D
Host Species Rabbit
Immunogen protein phosphatase 1, regulatory(inhibitor) subunit 14D. (Uniprot# Q9NXH3)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54866
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.