missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPP1R14C Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550996
This item is not returnable.
View return policy
Description
PP1R14C Polyclonal Antibody detects endogenous levels of PP1R14C protein.
Inhibitor of the PP1 regulatory subunit PPP1CA.
Specifications
| PPP1R14C | |
| Polyclonal | |
| Unconjugated | |
| PPP1R14C | |
| 6330514J04Rik; CPI17-like; Kepi; kinase C-enhanced PP1 inhibitor; kinase-enhanced PP1 inhibitor; NY-BR-81; PKC-potentiated PP1 inhibitory protein; Ppp1r14c; protein phosphatase 1 regulatory inhibitor subunit 14C; protein phosphatase 1 regulatory subunit 14C; protein phosphatase 1, regulatory (inhibitor) subunit 14C; serologically defined breast cancer antigen NY-BR-81 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 171010, 76142, 81706 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8R4R9, Q8R4S0, Q8TAE6 | |
| PPP1R14C | |
| Synthesized peptide derived from the Internal region of human KEPI. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction