missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R12B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13366-1-AP-150UL
This item is not returnable.
View return policy
Specifications
| PPP1R12B | |
| Polyclonal | |
| Unconjugated | |
| PPP1R12B | |
| MYPT2, PPP1R12B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 304813, 329251, 4660 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60237, Q8BG95 | |
| Ppp1r12b | |
| PPP1R12B Fusion Protein Ag4185 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction