missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPP1R11 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15857943
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15857943 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15857943 Supplier Invitrogen™ Supplier No. PA569534

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

This gene encodes a specific inhibitor of protein phosphatase-1 (PP1) with a differential sensitivity toward the metal-independent and metal-dependent forms of PP1. The gene is located within the major histocompatibility complex class I region on chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R11
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PPP1R11
Gene Accession No. O60927
Gene Alias 1500041B02Rik; AV117883; CFAP255; E3 ubiquitin-protein ligase PPP1R11; HCG V; HCGV; HCG-V; Hemochromatosis candidate gene V protein; hypothetical protein LOC550290; inhibitor-3; IPP3; Ppp1r11; protein phosphatase 1 regulatory inhibitor subunit 11; protein phosphatase 1 regulatory subunit 11; protein phosphatase 1, regulatory (inhibitor) subunit 11; protein phosphatase 1, regulatory (inhibitor) subunit 11, isoform 1; protein phosphatase inhibitor 3; T-complex testis expressed protein 5; t-complex testis expressed-5; t-complex testis-expressed 5; t-complex-associated-testis-expressed 5; TCTE5; Tctex5; Tctex-5; wu:fj64e04; zgc:110245
Gene Symbols PPP1R11
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human PPP1R11.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6992
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.