missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPP1R10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561475
This item is not returnable.
View return policy
Description
Immunogen sequence: TAPSHAKFRS TGLELETPSL VPVKKNASTV VVSDKYNLKP IPLKRQSNVA APGDATPPAE KKYKPLNTTP NATKEIKVKI IPPQPMEGLG FLDAL Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 94%.
RNA polymerases transcribe nuclear genes for ribosomal RNA, thus representing ribosomal biogenesis. RNA polymerase I (Pol I) is located in the nucleolus and transcribes class I genes, which code for large ribosomal RNA. Different subunits of the Pol I transcription machinery are targets of various physiological stimuli, which suggests that multiple signaling pathways are involved in carrying out Pol I transcription. RPA40 and RPA16 are subunits of Pol I that associate with each other at an early stage of RNA polymerase I assembly. RPA40 is essential for the function and integrity of the complex and is also an essential subunit of RNA polymerase III (Pol III). RPA40, RPA16 and RPA135 encode the three subunits of RNA polymerase I, respectively. RPA194 is the largest subunit of RNA Pol I and is not a component of Pol II and Pol III.
Specifications
| PPP1R10 | |
| Polyclonal | |
| Unconjugated | |
| PPP1R10 | |
| 2610025H06Rik; Cat53; D17Ertd808e; Fb19; HLA-C adjacent transcript 53; HLA-C associated transcript 53; MHC class I region proline-rich protein CAT53; p99; Phosphatase 1 nuclear targeting subunit; Pnuts; PNUTSPP1R10; PP1-binding protein of 114 kDa; PP1R10; PPP1R10; Protein FB19; protein phosphatase 1 regulatory subunit 10; protein phosphatase 1, regulatory (inhibitor) subunit 10; protein phosphatase 1, regulatory subunit 10; protein phosphatase I; Protein PNUTS; putative protein phosphatase 1 nuclear targeting subunit; R111; Serine/threonine-protein phosphatase 1 regulatory subunit 10; testis tissue sperm-binding protein Li 67n | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5514 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96QC0 | |
| PPP1R10 | |
| Recombinant protein corresponding to Human PPP1R10. Recombinant protein control fragment (Product #RP-103005). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction