missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1CC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11082-1-AP-20UL
This item is not returnable.
View return policy
Description
DYRK1B is a member of the DYRK family of protein kinases. DYRK1B contains a bipartite nuclear localization signal and is found mainly in muscle and testis. The protein is proposed to be involved in the regulation of nuclear functions. Three isoforms of DYRK1B have been identified differing in the presence of two alternatively spliced exons within the catalytic domain.Specifications
| PPP1CC | |
| Polyclonal | |
| Unconjugated | |
| PPP1CC | |
| PP1 Gamma, PP 1G, PPP1CC, PPP1G | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19047, 24669, 5501 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P36873, P63087 | |
| Ppp1cc | |
| PPP1CC Fusion Protein Ag1550 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction