missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPME1 Monoclonal Antibody (OTI10B2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525015
This item is not returnable.
View return policy
Description
PPME1 Monoclonal Antibody for Western Blot, ICC/IF, Flow
Protein phosphatase methylesterase-1 catalyzes the demethylation of the protein phosphatase-2A catalytic subunit.
Specifications
| PPME1 | |
| Monoclonal | |
| 0.87 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9Y570 | |
| Ppme1 | |
| Full length human recombinant protein of PPME1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| OTI10B2 | |
| Unconjugated | |
| Ppme1 | |
| 1110069N17Rik; 2700017M01Rik; Pme1; PME-1; PP2593; PPME1; PRO0750; Protein phosphatase methylesterase 1; protein phosphatase methylesterase-1; RGD1309683; testicular secretory protein Li 39 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 51400 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction