missing translation for 'onlineSavingsMsg'
Learn More

PPME1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£222.00 - £513.00

Specifications

Antigen PPME1
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18220764
View Documents
Novus Biologicals
NBP2-57103
100 μL
£513.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18686246
View Documents
Novus Biologicals
NBP2-57103-25ul
25 μL
£222.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PPME1 Polyclonal specifically detects PPME1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

PPME1
Polyclonal
Rabbit
Lipid and Metabolism
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
51400
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:AIAVHTASSNLVPSLLGLCMIDVVEGTAMDALNSMQNFLRGRPKTFKSLENAIEWSVKSGQIRNLESARVSMVGQVKQCEGITSPEGSKSIVEGIIEEE
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
Human
EC 3.1.1, EC 3.1.1.-, FLJ22226, PME-1PME1, protein phosphatase methylesterase 1, protein phosphatase methylesterase-1
PPME1
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.