missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPM1M Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94335-0.02ml
This item is not returnable.
View return policy
Description
PPM1M Polyclonal antibody specifically detects PPM1M in Human, Mouse samples. It is validated for Western Blot
Specifications
| PPM1M | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FLJ32332, PP2CEEC 3.1.3.16, PP2Ceta, PP2C-eta, PPM1E, protein phosphatase 1M, protein phosphatase 1M (PP2C domain containing), protein phosphatase 2C eta, protein phosphatase 2C eta-2, Protein phosphatase 2C isoform eta, protein phosphatase, Mg2+/Mn2+ dependent, 1M | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 213-459 of human PPM1M (NP_653242.3). MGGCTALVAVSLQGKLYMANAGDSRAILVRRDEIRPLSFEFTPETERQRIQQLAFVYPELLAGEFTRLEFPRRLKGDDLGQKVLFRDHHMSGWSYKRVEKSDLKYPLIHGQGRQARLLGTLAVSRGLGDHQLRVLDTNIQLKPFLLSVPQVTVLDVDQLELQEDDVVVMATDGLWDVLSNEQVAWLVRSFLPGNQEDPHRFSKLAQMLIHSTQGKEDSLTEEGQVSYDDVSVFVIPLHSQGQESSDH | |
| 0.02 mL | |
| Protein Phosphatase | |
| 132160 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction