missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPM1K Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14573-1-AP-150UL
This item is not returnable.
View return policy
Description
PPM1K regulates the mitochondrial permeability transition pore and is essential for cellular survival and development.Specifications
| PPM1K | |
| Polyclonal | |
| Unconjugated | |
| PPM1K | |
| PP2C kappa, PP2Cm, PPM1K, PTMP, UG0882E07 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 152926, 243382, 312381 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.62 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BXN7, Q8N3J5 | |
| Ppm1k | |
| PPM1K Fusion Protein Ag5660 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction