missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPM1K Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-85035
This item is not returnable.
View return policy
Description
PPM1K Polyclonal specifically detects PPM1K in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifikationer
| PPM1K | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml | |
| DKFZp667B084, DKFZp761G058, mitochondrial, PP2C domain-containing protein phosphatase 1K, PP2Ckappa, PP2C-like mitochondrial protein, PP2Cm, PP2C-type mitochondrial phosphoprotein phosphatase, protein phosphatase 2C kappa, protein phosphatase, Mg2+/Mn2+ dependent, 1K, PTMPEC 3.1.3.16 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| PPM1K | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:PETKRIKLHHADDSFLVLTTDGINFMVNSQEICDFVNQCHDPNEAAHAVTEQAIQYGTEDNSTAVVVPFGAWGKYKNSEINFSFS | |
| 0.1 mL | |
| Protein Phosphatase | |
| 152926 | |
| Human | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering