missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PPM1J Polyclonal antibody specifically detects PPM1J in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | PPM1J |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Formulation | PBS (pH 7.2) and 40% Glycerol |
| Gene Alias | DKFZp434P1514, FLJ35951, MGC19531, MGC90149, PP2CZ, PP2Czeta, PP2C-zeta, PPP2CZEC 3.1.3.16, protein phosphatase 1J, protein phosphatase 1J (PP2C domain containing), protein phosphatase 2a, catalytic subunit, zeta isoform, Protein phosphatase 2C isoform zeta, protein phosphatase 2C zeta, protein phosphatase, Mg2+/Mn2+ dependent, 1J |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against a recombinant protein corresponding to the amino acids: PEVRVYDLTQYEHCPDDVLVLGTDGLWDVTTDCEVAATVDRVLSAYEPNDHSRYTALAQALVLGARGTPRDRGWRLPNNKLGSGD |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?