missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPM1G Recombinant Rabbit Monoclonal Antibody (23GB2590), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | PPM1G |
|---|---|
| Clone | 23GB2590 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087402
![]() |
Invitrogen™
MA553538 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087769
|
Invitrogen™
MA553539 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PPM1G Recombinant Monoclonal antibody specifically detects PPM1G in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| PPM1G | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| F1LNI5, O15355, Q61074 | |
| 14208, 259229, 5496 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB2590 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AI385652; fibroblast growth factor inducible 13; fibroblast growth factor-inducible protein 13; Fin13; PP2C, gamma; PP2CG; PP2CGAMMA; PP2C-gamma; Ppm1c; Ppm1g; PPP2CG; protein phosphatase 1C; Protein phosphatase 1G; protein phosphatase 1G (formerly 2C), magnesium-dependent, gamma isoform; protein phosphatase 2, catalytic subunit, gamma isoform; protein phosphatase 2C gamma isoform; protein phosphatase 2C isoform gamma; Protein phosphatase magnesium-dependent 1 gamma; protein phosphatase, Mg2+/Mn2+ dependent 1G; protein phosphatase, Mg2+/Mn2+ dependent, 1G | |
| A synthesized peptide derived from human PPM1G (301-450AA). | |
| Primary | |
| -20°C | |
| Ppm1g |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title