Learn More
PPM1F Mouse, anti-Human, Clone: 2A2E8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68626-1-IG-150UL
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
The PPM1F antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PPM1F. This antibody recognizes human antigen. The PPM1F antibody has been validated for the following applications: WB, ELISA analysis.
Tekniske data
| PPM1F | |
| 2A2E8 | |
| CaM kinase phosphatase, CaMKPase, FEM 2, hFEM 2, KIAA0015, Partner of PIX 2, POPX2, PPM1F, Protein fem 2 homolog, Protein phosphatase 1F | |
| 150 μL | |
| 9647 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG2a |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion