Learn More
PPM1F Mouse, anti-Human, Clone: 2A2E8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68626-1-IG-20UL
This item is not returnable.
View return policy
Description
The PPM1F antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PPM1F. This antibody recognizes human antigen. The PPM1F antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| PPM1F | |
| 2A2E8 | |
| CaM kinase phosphatase, CaMKPase, FEM 2, hFEM 2, KIAA0015, Partner of PIX 2, POPX2, PPM1F, Protein fem 2 homolog, Protein phosphatase 1F | |
| 20 μL | |
| 9647 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG2a |