Learn More
PPM1E Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25089-1-AP-150UL
Description
The PPM1E antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PPM1E. This antibody recognizes human, mouse, rat antigen. The PPM1E antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| PPM1E | |
| 900 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, Immunofluorescence, Immunohistochemistry, ELISA | |
| CaMKN, CaMKP N, CaMKP nucleus, KIAA1072, Partner of PIX 1, Partner of PIX alpha, Partner of PIXA, POPX1, PP2CH, PPM1E, Protein phosphatase 1E | |
| 150 μL | |
| 22843 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |