missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPM1E Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104040
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PPM1E.
Protein phosphatase that inactivates multifunctional CaM kinases such as CAMK4 and CAMK2. Dephosphorylates and inactivates PAK. May play a role in the inhibition of actin fiber stress breakdown and in morphological changes driven by TNK2/CDC42. Dephosphorylates PRKAA2.
Specifications
| PPM1E | |
| Polyclonal | |
| Unconjugated | |
| PPM1E | |
| AW049266; B930008A12Rik; Ca(2+)/calmodulin-dependent protein kinase phosphatase N; calmodulin-dependent protein kinase phosphatase N; caMKN; CaMKP-N; caMKP-nucleus; Kiaa1072; mKIAA1072; nuclear calmodulin-dependent protein kinase phosphatase; partner of PIX 1; partner of PIXA; Partner of PIX-alpha; POPX1; PP2CE; PP2Ceta; PP2C-eta; PP2CH; PPM1E; PPM1M; Protein phosphatase 1E; protein phosphatase 1E (PP2C domain containing); protein phosphatase 1M; protein phosphatase 1M (PP2C domain containing); protein phosphatase 2C eta-2; protein phosphatase 2C isoform eta; protein phosphatase, Mg2+/Mn2+ dependent 1E; protein phosphatase, Mg2+/Mn2+ dependent 1M; protein phosphatase, Mg2+/Mn2+ dependent, 1E; protein phosphatase, Mg2+/Mn2+ dependent, 1M | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 132160, 22843, 320472, 360593, 67905, 686298 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q80TL0, Q80Z30, Q8BU27, Q8WY54, Q96MI6 | |
| PPM1E, PPM1M | |
| A synthesized peptide derived from human PPM1E(Accession Q8WY54), corresponding to amino acid residues A475-C525. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction