missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPM1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582557
This item is not returnable.
View return policy
Description
Immunogen sequence: KRNVIEAVYS RLNPHRESDG ASDEAEESGS QGKLVEALRQ MRINHRGNYR QLLEEMLTSY RLAKVEGEES PAEPAATATS SNSDAGNPVT MQESHTESES GLAEL Highest antigen sequence indentity to the following orthologs: Mouse - 80%, Rat - 85%.
The protein encoded by this gene is a member of the PP2C family of Ser/Thr protein phosphatases. PP2C family members are known to be negative regulators of cell stress response pathways. This phosphatase has been shown to dephosphorylate cyclin-dependent kinases (CDKs), and thus may be involved in cell cycle control. Overexpression of this phosphatase is reported to cause cell-growth arrest or cell death. Alternative splicing results in multiple transcript variants encoding different isoforms. Additional transcript variants have been described, but currently do not represent full-length sequences.
Specifications
| PPM1B | |
| Polyclonal | |
| Unconjugated | |
| PPM1B | |
| EGK_05287; Pp2c2; PP2CB; PP2CBETA; PP2C-beta; PP2C-beta-X; PPC2BETAX; PPM1B; Pppm1b; Protein phosphatase 1B; protein phosphatase 1B (formerly 2C), magnesium-dependent, beta isoform; protein phosphatase 1B, magnesium dependent, beta isoform; protein phosphatase 2C isoform beta; protein phosphatase 2C-like protein; Protein phosphatase type 1B (formely 2C) Mg-dependent beta isoform; Protein phosphatase type 1B (formely 2C), Mg-dependent, beta isoform; protein phosphatase, Mg2+/Mn2+ dependent 1B; protein phosphatase, Mg2+/Mn2+ dependent, 1B; Ser/Thr protein phosphatase type 2C beta 2 isoform | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5495 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75688 | |
| PPM1B | |
| Recombinant protein corresponding to Human PPM1B. Recombinant protein control fragment (Product #RP-91561). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction