Learn More
PPM1B Mouse anti-Human, Unconjugated, Clone: 2C7H2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67647-1-IG-150UL
This item is not returnable.
View return policy
Description
The PPM1B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PPM1B. This antibody recognizes human antigen. The PPM1B antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| PPM1B | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Immunohistochemistry, ELISA | |
| 2C7H2 | |
| Unconjugated | |
| 150 μL | |
| 5495 | |
| IgG1 |