missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPIP5K2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558390
This item is not returnable.
View return policy
Description
Immunogen sequence: LSVSSPEGTG TWLHYTSGVG TGRRRRRSGE QITSSPVSPK SLAFTSSIFG SWQQVVSENA NYLRTPRTLV EQKQNPTVGS HCA Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 31%.
PPIP5K2, also known as inositol hexakisphosphate and diphosphoinositol pentakisphosphate kinase 2, is a gene located on chromosome 5 in humans. This gene encodes a kinase involved in the phosphorylation of inositol pyrophosphates, which are essential for cellular signaling and regulation processes. Specifically, PPIP5K2 phosphorylates InsP6 and 1-InsP7 at the 5 position, playing a significant role in both cellular proliferation and metastasis. Recent studies have shown that PPIP5K2 can facilitate the growth and spread of non-small lung cancer (NSCLC) and colorectal carcinoma. It promotes cellular proliferation and migration independent of its kinase activity, in part by modifying its interaction with proteins like 14-3-3, affecting its subcellular distribution. Structural analysis of PPIP5K2 reveals that it possesses unique kinetic properties distinct from other inositol kinase isoforms. Genetic variants, such as the Arg837His missense variant, can alter its phosphatase activity, potentially impacting its functionality in signaling pathways. PPIP5K2 is intricately involved in epithelial-mesenchymal transition (EMT) mechanisms, further influencing cancer cell metastasis. Understanding the biochemical pathways and genetic variations associated with PPIP5K2 is crucial in comprehending its role in disease pathogenesis and therapeutic targeting.
Specifications
| PPIP5K2 | |
| Polyclonal | |
| Unconjugated | |
| PPIP5K2 | |
| AW555814; CFAP160; D330021B20; diphosphoinositol pentakisphosphate kinase 2; Hisppd1; histidine acid phosphatase domain containing 1; histidine acid phosphatase domain-containing protein 1; hsVIP2; inositol heptaphosphate kinase 2; inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 2; InsP6 and PP-IP5 kinase 2; IP7K2; KIAA0433; LOW QUALITY PROTEIN: inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 2; mKIAA0433; mmVIP2; PPIP5K2; VIP1 homolog 2; Vip2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23262 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43314 | |
| PPIP5K2 | |
| Recombinant protein corresponding to Human PPIP5K2. Recombinant protein control fragment (Product #RP-96885). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction