missing translation for 'onlineSavingsMsg'
Learn More

PPIL2 Antibody, FITC, Novus Biologicals™

Product Code. 18364252 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18364252 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18364252 Supplier Novus Biologicals Supplier No. NBP300291F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PPIL2 Polyclonal specifically detects PPIL2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPIL2
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias CYC4, cyclophilin, 60kDa, cyclophilin-60, Cyclophilin-like protein Cyp-60, Cyp-60, CYP60, EC 5.2.1.8, FLJ39930, hCyP-60, MGC33174, MGC787, peptidylprolyl cis-trans isomerase, peptidyl-prolyl cis-trans isomerase-like 2, peptidylprolyl isomerase (cyclophilin)-like 2, PPIase, Rotamase PPIL2
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant PPIL2 (Q13356-2; Gly280-Pro457).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23759
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.