missing translation for 'onlineSavingsMsg'
Learn More

PPIL2 Antibody (2A10), Novus Biologicals™

Product Code. 18374819 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18374819 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18374819 Supplier Novus Biologicals Supplier No. H00023759M04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PPIL2 Monoclonal antibody specifically detects PPIL2 in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPIL2
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2A10
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH00022
Gene Alias CYC4, cyclophilin, 60kDa, cyclophilin-60, Cyclophilin-like protein Cyp-60, Cyp-60, CYP60, EC 5.2.1.8, FLJ39930, hCyP-60, MGC33174, MGC787, peptidylprolyl cis-trans isomerase, peptidyl-prolyl cis-trans isomerase-like 2, peptidylprolyl isomerase (cyclophilin)-like 2, PPIase, Rotamase PPIL2
Host Species Mouse
Immunogen PPIL2 (AAH00022, 1 a.a. ~ 527 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MGKRQHQKDKMYITCAEYTHFYGGKKPDLPQTNFRRLPFDHCSLSLQPFVYPVCTPDGIVFDLLNIVPWLKKYGTNPSNGEKLDGRSLIKLNFSKNSEGKYHCPVLFTVFTNNTHIVAVRTTGNVYAYEAVEQLNIKAKNFRDLLTDEPFSRQDIITLQDPTNLDKFNVSNFYHVKNNMKIIDPDEEKAKQDPSYYLKNTNAETRETLQELYKEFKGDEILAATMKAPEKKKVDKLNAAHYSTGKVSASFTSTAMVPETTHEAAAIDEDVLRYQFVKKKGYVRLHTNKGDLNLELHCDLTPKTCENFIRLCKKHYYDGTIFHRSIRNFVIQGGDPTGTGTGGESYWGKPFKDEFRPNLSHTGRGILSMANSGPNSNRSQFFITFRSCAYLDKKHTIFGRVVGGFDVLTAMENVESDPKTDRPKEEIRIDATTVFVDPYEEADAQIAQERKTQLKVAPETKVKSSQPQAGSQGPQTFRQGVGKYINPAATEQQRKSPQPVPLSPCPRRSPVGVLGTSAPGSSRLPDDH
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23759
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.