missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPIL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15144-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene is a member of the cyclophilin family of peptidylprolyl isomerases (PPIases). The cyclophilins are a highly conserved, ubiquitous family, members of which play an important role in protein folding, immunosuppression by cyclosporin A, and infection of HIV-1 virions. Based on similarity to other PPIases, this protein could accelerate the folding of proteins and might catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides.Specifications
| PPIL1 | |
| Polyclonal | |
| Unconjugated | |
| PPIL1 | |
| CGI 124, CYPL1, hCyPX, PPIase, PPIL1, Rotamase PPIL1, UNQ2425/PRO4984 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 309651, 51645, 68816 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 1.05 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9D0W5, Q9Y3C6 | |
| Ppil1 | |
| PPIL1 Fusion Protein Ag7436 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction