missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPIH Monoclonal Antibody (OTI1C6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526600
This item is not returnable.
View return policy
Description
PPIH Monoclonal Antibody for Western Blot
The protein encoded by this gene is a member of the peptidyl-prolyl cis-trans isomerase (PPIase) family. PPIases catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerate the folding of proteins. This protein is a specific component of the complex that includes pre-mRNA processing factors PRPF3, PRPF4, and PRPF18, as well as U4/U5/U6 tri-snRNP. This protein has been shown to possess PPIase activity and may act as a protein chaperone that mediates the interactions between different proteins inside the spliceosome.
Specifications
| PPIH | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43447 | |
| PPIH | |
| Full length human recombinant protein of PPIH produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1C6 | |
| Unconjugated | |
| PPIH | |
| 1100001J08Rik; 2010111B15Rik; 4833408F11Rik; AI464484; cyclophilin H; CYP20; CYP-20; CYPH; D4Wsu43e; peptidyl prolyl isomerase H; peptidyl-prolyl cis-trans isomerase H; peptidylprolyl isomerase H; peptidylprolyl isomerase H (cyclophilin H); PPIase h; Ppih; RGD1564921; Rotamase H; small nuclear ribonucleoprotein particle-specific cyclophilin H; SnuCyp-20; USA-CYP; USA-CyP SnuCyp-20; U-snRNP-associated cyclophilin SnuCyp-20; U-snRNP-associated cyclophilin SunCyp-20 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10465 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction