missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPIG Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12985-1-AP-20UL
This item is not returnable.
View return policy
Description
Structural Maintece of Chromosomes (SMC) family proteins play critical roles in various nuclear events that require structural changes of chromosomes, including mitotic chromosome organization, DNA recombination and repair and global transcriptional repression. The chromo proteins are conserved in eukaryotes and lead to mitotic chromosome segregatior defects. This suggests a critical function of SMC family proteins in mitotic chromosome dynamics. SMC1 and SMC3 form a heterodimeric complex required for metaphase progression in mitotic cells. Specifically this SMC1/SMC3 complex is responsible for sister chromatid cohesion during metaphase. Mutations in the gene can lead to Cornelia de Lange syndrome 2.Specifications
| PPIG | |
| Polyclonal | |
| Unconjugated | |
| PPIG | |
| CARS cyclophilin, CARS Cyp, CASP10, Clk associating RS cyclophilin, Cyclophilin G, CYP, Peptidyl prolyl isomerase G, PPIase G, PPIG, Rotamase G, SR cyclophilin, SR cyp, SRCyp | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 228005, 9360 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| A2AR02, Q13427 | |
| PPIG | |
| PPIG Fusion Protein Ag3622 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction