missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPFIBP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13953-1-AP-20UL
This item is not returnable.
View return policy
Specifications
| PPFIBP2 | |
| Polyclonal | |
| Unconjugated | |
| PPFIBP2 | |
| Cclp1, Liprin beta 2, PPFIBP2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19024, 308918, 8495 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O35711, Q8ND30 | |
| Ppfibp2 | |
| PPFIBP2 Fusion Protein Ag5064 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction