missing translation for 'onlineSavingsMsg'
Learn More

PPFIBP2 Antibody, Novus Biologicals™

Product Code. 18389869 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389869 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18389869 Supplier Novus Biologicals Supplier No. H00008495D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PPFIBP2 Polyclonal antibody specifically detects PPFIBP2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPFIBP2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_003612.1
Gene Alias Cclp1, DKFZp781K06126, liprin-beta-2, MGC42541, Protein tyrosine phosphatase receptor type f polypeptide-interactingprotein-binding protein 2, PTPRF interacting protein, binding protein 2 (liprin beta 2), PTPRF-interacting protein-binding protein 2
Host Species Rabbit
Immunogen PPFIBP2 (NP_003612.1, 1 a.a. - 876 a.a.) full-length human protein. MASDASHALEAALEQMDGIIAGTKTGADLSDGTCEPGLASPASYMNPFPVLHLIEDLRLALEMLELPQERAALLSQIPGPTAAYIKEWFEESLSQVNHHSAASNETYQERLARLEGDKESLILQVSVITDQVEAQGEKIRDLEVCLEGHQVKLNAAEEMLQQELLSRTSLETQKLDLMTEVSELKLKLVGMEKEQREQEEKQRKAEELLQELRHLKIKVEELENERNQYEWKLKATKAEVAQLQEQVALKDAEIERLHSQLSRTAALHSESHTERDQEIQRLKMGMETLLLANEDKDRRIEELTGLLNQYRKVKEIVMVTQGPSERTLSINEEEPEGGFSKWNATNKDPEELFKQEMPPRCSSPTVGPPPLPQKSLETRAQKKLSCSLEDLRSESVDKCMDGNQPFPVLEPKDSPFLAEHKYPTLPGKLSGATPNGEAAKSPPTICQPDATGSSLLRLRDTESGWDDTAVVNDLSSTSSGTESGPQSPLTPDGKRNPKGIKKFWGKIRRTQSGNFYTDTLGMAEFRRGGLRATAGPRLSRTRDSKGQKSDANAPFAQWSTERVCAWLEDFGLAQYVIFARQWVSSGHTLLTATPQDMEKELGIKHPLHRKKLVLAVKAINTKQEEKSALLDHIWVTRWLDDIGLPQYKDQFHESRVDGRMLQYLTVNDLLFLKVTSQLHHLSIKCAIHVLHVNKFNPHCLHRRPADESNLSPSEVVQWSNHRVMEWLRSVDLAEYAPNLRGSGVHGGLIILEPRFTGDTLAMLLNIPPQKTLLRRHLTTKFNALIGPEAEQEKREKMASPAYTPLTTTAKVRPRKLGFSHFGNIRKKKFDESTDYICPMEPSDGVSDSHRVYSGYRGLSPLDAPELDGLDQVGQIS
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8495
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.