missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPFIA4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-88078
This item is not returnable.
View return policy
Description
PPFIA4 Polyclonal specifically detects PPFIA4 in Human samples. It is validated for Western Blot.
Specifications
| PPFIA4 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| KIAA0897, liprin alpha4, liprin-alpha-4, Liprin-alpha4, Protein tyrosine phosphatase receptor type f polypeptide-interacting proteinalpha-4, protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interactingprotein (liprin), alpha 4, PTPRF-interacting protein alpha-4 | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of Human PPFIA4 | |
| 100 μL | |
| Protein Phosphatase | |
| 8497 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction