missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PPFIA3 Polyclonal antibody specifically detects PPFIA3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Immunogen | PPFIA3 antibody was raised against a 17 amino acid peptide near the carboxy terminus of human PPFIA3. The immunogen is located within amino acids 1100 - 1150 of PPFIA3. Amino Acid Squence: LGTDRRLDEDSAKSFSR |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?