missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPFIA1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119372
This item is not returnable.
View return policy
Description
PPFIA1 Polyclonal Antibody for Western Blot, ICC/IF
PPFIA1 may regulate the disassembly of focal adhesions. May localize receptor-like tyrosine phosphatases type 2A at specific sites on the plasma membrane, possibly regulating their interaction with the extracellular environment and their association with substrates.
Specifications
| PPFIA1 | |
| Polyclonal | |
| Unconjugated | |
| Ppfia1 | |
| C030014K08Rik; C87158; FLJ41337; FLJ42630; FLJ43474; LAR-interacting protein 1; LIP.1; LIP1; LIP-1; LIPRIN; Liprin-alpha1; liprin-alpha-1; MGC26800; PPFIA1; Protein tyrosine phosphatase receptor type f polypeptide-interacting protein alpha-1; protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein (liprin), alpha 1; protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein, alpha 1; PTPRF interacting protein alpha 1; PTPRF-interacting protein alpha-1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 8500 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q13136 | |
| Ppfia1 | |
| Recombinant protein Tyrosine Phosphatase F Interacting Protein 1. The antigen corresponds to amino acid range 1-250 of the target protein. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction