missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PPEF2 Polyclonal specifically detects PPEF2 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | PPEF2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | protein phosphatase 7, catalytic subunit, beta isozyme, protein phosphatase with EF hands 2, protein phosphatase, EF hand calcium-binding domain 2, protein phosphatase, EF-hand calcium binding domain 2, serine/threonine-protein phosphatase with EF-hands 2 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human PPEF2 (NP_006230). Peptide sequence LVTGEKEEPSRSASEADSEAGELRKPTQEEWRQVVDILWSDPMAQEGCKA |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?