missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPCS Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531620
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T. Predicted reactivity: Mouse (85%), Rat (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Biosynthesis of coenzyme A (CoA) from pantothenic acid (vitamin B5) is an essential universal pathway in prokaryotes and eukaryotes. PPCS (EC 6.3.2.5), one of the last enzymes in this pathway, converts phosphopantothenate to phosphopantothenoylcysteine.
Specifications
| PPCS | |
| Polyclonal | |
| Unconjugated | |
| PPCS | |
| 6330579B17Rik; AI413471; AI790434; Coab; Phosphopantothenate--cysteine ligase; phosphopantothenoylcysteine synthetase; PPC synthetase; Ppcs; RGD1311399 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79717 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.69 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9HAB8 | |
| PPCS | |
| Recombinant protein encompassing a sequence within the center region of human PPCS. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction