missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-PPCC oxalate, Tocris Bioscience™
Selective sigma (σ) agonist (σ1> σ2)
£172.64 - £652.93
Chemical Identifiers
CAS | 932736-91-9 |
---|---|
Molecular Formula | C52H62N2O14 |
Molecular Weight (g/mol) | 939.068 |
InChI Key | IGJDSEGFEVKASG-KLELIYFTSA-N |
Synonym | (S™, R™)-2-[(4-Hydroxy-4-phenyl-1-piperidinyl)methyl]-1-(4-methylphenyl)-cyclopropanecarboxylic acid methyl ester |
PubChem CID | 90488889 |
IUPAC Name | methyl (1S,2R)-2-[(4-hydroxy-4-phenylpiperidin-1-yl)methyl]-1-(4-methylphenyl)cyclopropane-1-carboxylate;methyl (1R,2S)-2-[(4-hydroxy-4-phenylpiperidin-1-yl)methyl]-1-(4-methylphenyl)cyclopropane-1-carboxylate;oxalic acid |
SMILES | CC1=CC=C(C=C1)C2(CC2CN3CCC(CC3)(C4=CC=CC=C4)O)C(=O)OC.CC1=CC=C(C=C1)C2(CC2CN3CCC(CC3)(C4=CC=CC=C4)O)C(=O)OC.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18065902
|
Tocris Bioscience
3870/10 |
10 mg |
£172.64
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18013852
|
Tocris Bioscience
3870/50 |
50 mg |
£652.93
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Selective sigma (σ) receptor ligand. Displays high affinity for σ1; also binds at σ2 sites (Ki = 1.5 nM and 50.8 nM respectively). Selective over a range of receptor types including dopaminergic and muscarinic receptors, DAT and SERT.Chemical Identifiers
932736-91-9 | |
939.068 | |
(S™, R™)-2-[(4-Hydroxy-4-phenyl-1-piperidinyl)methyl]-1-(4-methylphenyl)-cyclopropanecarboxylic acid methyl ester | |
methyl (1S,2R)-2-[(4-hydroxy-4-phenylpiperidin-1-yl)methyl]-1-(4-methylphenyl)cyclopropane-1-carboxylate;methyl (1R,2S)-2-[(4-hydroxy-4-phenylpiperidin-1-yl)methyl]-1-(4-methylphenyl)cyclopropane-1-carboxylate;oxalic acid |
C52H62N2O14 | |
IGJDSEGFEVKASG-KLELIYFTSA-N | |
90488889 | |
CC1=CC=C(C=C1)C2(CC2CN3CCC(CC3)(C4=CC=CC=C4)O)C(=O)OC.CC1=CC=C(C=C1)C2(CC2CN3CCC(CC3)(C4=CC=CC=C4)O)C(=O)OC.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O |
Specifications
932736-91-9 | |
(S™, R™)-2-[(4-Hydroxy-4-phenyl-1-piperidinyl)methyl]-1-(4-methylphenyl)-cyclopropanecarboxylic acid methyl ester | |
CC1=CC=C(C=C1)C2(CC2CN3CCC(CC3)(C4=CC=CC=C4)O)C(=O)OC.CC1=CC=C(C=C1)C2(CC2CN3CCC(CC3)(C4=CC=CC=C4)O)C(=O)OC.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O | |
939.068 | |
469.53 | |
(±)-PPCC oxalate |
C52H62N2O14 | |
IGJDSEGFEVKASG-KLELIYFTSA-N | |
methyl (1S,2R)-2-[(4-hydroxy-4-phenylpiperidin-1-yl)methyl]-1-(4-methylphenyl)cyclopropane-1-carboxylate;methyl (1R,2S)-2-[(4-hydroxy-4-phenylpiperidin-1-yl)methyl]-1-(4-methylphenyl)cyclopropane-1-carboxylate;oxalic acid | |
90488889 | |
>99% |
Safety and Handling
Recommended Storage : Store at +4°C