Learn More
PeproTech PPBP Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Brand: PeproTech 500-P03-1MG
Description
AA Sequence of recombinant protein: AELRCMCIKT TSGIHPKNIQ SLEVIGKGTH CNQVEVIATL KDGRKICLDP DAPRIKKIVQ KKLAGDESAD. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human NAP-2 (CXCL7). Anti-Human NAP-2 (CXCL7)-specific antibody was purified by affinity chromatography employing an immobilized Human NAP-2 matrix. Sandwich ELISA: To detect Human NAP-2 (CXCL7) by sandwich ELISA (using 100 μL/well) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human NAP-2 (CXCL7) (500-P03BT) as a detection antibody, allows the detection of at least 2000- 4000 pg/mL of Recombinant Human NAP-2 (CXCL7). Western Blot: To detect Human NAP-2 (CXCL7) by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents the detection limit for Recombinant Human NAP-2 (CXCL7) is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Members of the a-chemokine subfamily of inducible, secreted, pro-inflammatory cytokines contain a similar motif, in which the first two cysteine residues are separated by a single residue (Cys-X-Cys), and are also chemotactic for neutrophils. The platelet basic protein (PBP), a member of the a-chemokine family, resides in the a-granules of platelets and is released upon their activation. Proteolytic cleavage of the amino terminus of PBP leads to the generation of several peptides, which include mature PBP, connective tissue-activating peptide III (CTAP III, also designated low affinity platelet factor IV (LA-PF4)), b-thromboglobulin (b-TG), and neutrophil-activating peptide 2 (NAP-2). PBP and its N-truncated derivatives mediate inflammation and wound healing. Specifically, NAP-2 activates chemotaxis and degranulation in neutrophils during inflammation. The gene encoding human PBP maps to chromosome 4q12-q13.
Specifications
| PPBP | |
| Polyclonal | |
| Unconjugated | |
| PPBP | |
| 2400003M24Rik; AI854500; Beta-TG; Beta-thromboglobulin; b-TG1; chemokine (C-X-C motif) ligand 7; connective tissue-activating peptide III; Connective tissue-activating peptide III(1-81); CTAP3; CTAPIII; CTAP-III; CTAP-III(1-81); CXC chemokine ligand 7; C-X-C motif chemokine 7; CXCL7; LA-PF4; LDGF; Leukocyte-derived growth factor; low-affinity platelet factor IV; Macrophage-derived growth factor; MDGF; NAP-2; NAP-2(1-63); NAP-2(1-66); NAP-2(73); NAP-2(74); NAP-2-L1; neutrophil activating peptide-2; neutrophil-activating peptide 2; Neutrophil-activating peptide 2(1-63); Neutrophil-activating peptide 2(1-66); Neutrophil-activating peptide 2(73); Neutrophil-activating peptide 2(74); neutrophil-activating peptide-2; PBP; Platelet basic protein; PPBP; pro-platelet basic protein; pro-platelet basic protein (chemokine (C-X-C motif) ligand 7); SCYB7; small inducible cytokine B7; small inducible cytokine subfamily B, member 7; small-inducible cytokine B7; TC1; TC-1; TC2; TC-2; TGB; TGB1; THBGB; THBGB1; thrombocidin 1; thrombocidin 2; thromboglobulin, beta-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5473 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P02775 | |
| PPBP | |
| E.coli-derived Recombinant Human NAP-2 (CXCL7). | |
| 1 mg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.