missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ PPARG Recombinant Human Protein, N-His.GST tag

Product Code. 15285685
Change view
Click to view available options
Quantity:
5 μg
Unit Size:
5µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15285685 5 μg 5µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15285685 Supplier Sino Biological™ Supplier No. 12019H20B5

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Expressed from DNA sequence encoding human PPARG isoform 2 (Met 1-Tyr 505) fused to N-terminal polyhistidine-tagged GST tag at N-Terminus

  • Formulation: 50mM Tris, 100mM NaCl, 20% Glycerol, 0.3 mM DTT, pH 7.4
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Protein Structure Analysis, Assay Standard
Gene ID (Entrez) 5468
Molecular Weight (g/mol) 85.4kd
Quantity 5 μg
Source Human
Structural Form Full Length, Recombinant
Endotoxin Concentration <1 EU/μg
Common Name Peroxisome Proliferator-Activated Receptor Gamma
Gene Symbol PPARG
Product Type PPARG Recombinant Human Protein
Protein Tag GST Tag, His Tag
Expression System Insect
Protein Family Other proteins
Form Lyophilized
Purity or Quality Grade ≥90% by SDS-PAGE
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.