missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPARD Mouse anti-Human, Mouse, Clone: 1B10E1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60193-1-IG-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the peroxisome proliferator-activated receptor (PPAR) family. PPARs are nuclear hormone receptors that bind peroxisome proliferators and control the size and number of peroxisomes produced by cells. PPARs mediate a variety of biological processes, and may be involved in the development of several chronic diseases, including diabetes, obesity, atherosclerosis, and cancer. This protein is a potent inhibitor of ligand-induced transcription activity of PPAR alpha and PPAR gamma. It may function as an integrator of transcription repression and nuclear receptor signaling. The expression of this gene is found to be elevated in colorectal cancer cells. The elevated expression can be repressed by adenomatosis polyposis coli (APC), a tumor suppressor protein related to APC/beta-catenin signaling pathway. Knockout studies in mice suggested the role of this protein in myelination of the corpus callosum, lipid metabolism, and epidermal cell proliferation.Specifications
| PPARD | |
| Monoclonal | |
| 0.6 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P35396, Q03181 | |
| Ppard | |
| PPARD Fusion Protein Ag16770 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 1B10E1 | |
| Unconjugated | |
| PPARD | |
| FAAR, NR1C2, NUC1, NUCI, NUCII, Nuclear hormone receptor 1, PPAR beta, PPAR delta, PPARB, PPARD | |
| Mouse | |
| Protein A | |
| RUO | |
| 19015, 5467 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction