missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPAR gamma Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16643-1-AP-20UL
This item is not returnable.
View return policy
Description
PPP1R10 is a protein with similarity to a rat protein that has an inhibitory effect on protein phosphatase-1 (PP1). The rat protein localizes to the nucleus and colocalizes with chromatin at distinct phases during mitosis. This gene encodes a protein with similarity to a rat protein that has an inhibitory effect on protein phosphatase-1 (PP1). The rat protein localizes to the nucleus and colocalizes with chromatin at distinct phases during mitosis. This gene lies within the major histocompatibility complex class I region on chromosome 6.Specifications
| PPAR gamma | |
| Polyclonal | |
| Unconjugated | |
| PPARG | |
| CIMT1, NR1C3, PPAR gamma, PPARG, PPARG1, PPARG2, PPARgamma | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19016, 25664, 5468 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O88275, P37231, P37238 | |
| Pparg | |
| PPAR gamma Fusion Protein Ag10005 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction