missing translation for 'onlineSavingsMsg'
Learn More

PPAR gamma/NR1C3 Rabbit anti-Rat, Clone: 9P1J6, Novus Biologicals™

Product Code. 18369606
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18369606 100 μg 100µL
18349793 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18369606 Supplier Novus Biologicals Supplier No. NBP315853100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PPAR gamma/NR1C3 Monoclonal antibody specifically detects PPAR gamma/NR1C3 in Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPAR gamma/NR1C3
Applications Western Blot
Classification Monoclonal
Clone 9P1J6
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias CIMT1, NR1C3GLM1, Nuclear receptor subfamily 1 group C member 3, peroxisome proliferator-activated receptor gamma, peroxisome proliferator-activated receptor gamma 1, PPAR gamma, PPARG1peroxisome proliferative activated receptor, gamma, PPARG2PPARgamma, PPAR-gamma
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 50-150 of human PPAR gamma/NR1C3 (P37231).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5468
Target Species Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.