missing translation for 'onlineSavingsMsg'
Learn More

PPAR gamma/NR1C3 Antibody, DyLight 488, Novus Biologicals™

Product Code. 18418306
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418306 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418306 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP222106G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PPAR gamma/NR1C3 Polyclonal antibody specifically detects PPAR gamma/NR1C3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPAR gamma/NR1C3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias CIMT1, NR1C3GLM1, Nuclear receptor subfamily 1 group C member 3, peroxisome proliferator-activated receptor gamma, peroxisome proliferator-activated receptor gamma 1, PPAR gamma, PPARG1peroxisome proliferative activated receptor, gamma, PPARG2PPARgamma, PPAR-gamma
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human PPAR gamma protein (between residues 20-120) [UniProt P37231]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5468
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.