missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPAR gamma Mouse anti-Human, Clone: 4E12F10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60127-1-IG-20UL
This item is not returnable.
View return policy
Description
PPP1R10 is a protein with similarity to a rat protein that has an inhibitory effect on protein phosphatase-1 (PP1). The rat protein localizes to the nucleus and colocalizes with chromatin at distinct phases during mitosis. This gene encodes a protein with similarity to a rat protein that has an inhibitory effect on protein phosphatase-1 (PP1). The rat protein localizes to the nucleus and colocalizes with chromatin at distinct phases during mitosis. This gene lies within the major histocompatibility complex class I region on chromosome 6.Specifications
| PPAR gamma | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P37231 | |
| PPARG | |
| PPAR gamma Fusion Protein Ag10005 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot | |
| 4E12F10 | |
| Unconjugated | |
| PPARG | |
| CIMT1, NR1C3, PPAR gamma, PPARG, PPARG1 | |
| Mouse | |
| Caprylic Acid Precipitation | |
| RUO | |
| 5468 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction