missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPAR delta/NR1C2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22468-0.025ml
This item is not returnable.
View return policy
Description
PPAR delta/NR1C2 Polyclonal specifically detects PPAR delta/NR1C2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| PPAR delta/NR1C2 | |
| Polyclonal | |
| Unconjugated | |
| 1 mg/mL BSA with 0.05% Sodium Azide | |
| PPARD | |
| Synthetic peptide corresponding to residues M(1) E Q P Q E E T P E A R E E(14) C of mouse PPAR delta. | |
| 0.025 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| 1 mg/mL | |
| Western Blot 1:250 - 1:750, Flow Cytometry 3 - 5 ug 106 cells, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 2 μg/mL, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:200 | |
| FAAR, MGC3931, NR1C2PPAR-delta, NUC1PPAR-beta, NUCI, NUCII, Nuclear hormone receptor 1, Nuclear receptor subfamily 1 group C member 2, peroxisome proliferative activated receptor, delta, Peroxisome proliferator-activated receptor beta, peroxisome proliferator-activated receptor delta, PPARB | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 5467 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction