missing translation for 'onlineSavingsMsg'
Learn More

PPAR delta/NR1C2 Antibody (4E3-1B11), Novus Biologicals™

Product Code. 18371529 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18371529 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18371529 Supplier Novus Biologicals Supplier No. H00005467M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PPAR delta/NR1C2 Monoclonal antibody specifically detects PPAR delta/NR1C2 in Human samples. It is validated for Western Blot, ELISA, Proximity Ligation Assay, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPAR delta/NR1C2
Applications Western Blot, ELISA, Proximity Ligation Assay, Sandwich ELISA
Classification Monoclonal
Clone 4E3-1B11
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Proximity Ligation Assay, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH02715
Gene Alias FAAR, MGC3931, NR1C2PPAR-delta, NUC1PPAR-beta, NUCI, NUCII, Nuclear hormone receptor 1, Nuclear receptor subfamily 1 group C member 2, peroxisome proliferative activated receptor, delta, Peroxisome proliferator-activated receptor beta, peroxisome proliferator-activated receptor delta, PPARB
Host Species Mouse
Immunogen PPARD (AAH02715, 1 a.a. ~ 361 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MEQPQEEAPEVREEEEKEEVAEAEGAPELNGGPQHALPSSSYTDLSRSSSPPSLLDQLQMGCDGASCGSLNMECRVCGDKASGFHYGVHACEGCKGFFRRTIRMKLEYEKCERSCKIQKKNRNKCQYCRFQKCLALGMSHNAIRFGRMPEAEKRKLVAGLTANEGSQYNPQVADLKAFSKHIYNAYLKNFNMTKKKARSILTGKASHTAPFVIHDIETLWQAEKGLVWKQLVNGLPPYKEISVHVFYRCQCTTVETVRELTEFAKSIPSFSSLFLNDQVTLLKYGVHEAIFAMLASIVNKDGLLVANGSGFVTREFLRSLRKPFSDIIEPKFEFAVKFNALELDDSDLALFIAAIILCGGE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Transcription Factors and Regulators, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5467
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.