missing translation for 'onlineSavingsMsg'
Learn More

PPAR alpha/NR1C1 Antibody (OTI1E8), DyLight 594, Novus Biologicals™

Product Code. 18404837 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18404837 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18404837 Supplier Novus Biologicals Supplier No. NBP273567DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PPAR alpha/NR1C1 Monoclonal antibody specifically detects PPAR alpha/NR1C1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPAR alpha/NR1C1
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone OTI1E8
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias alpha, hPPAR, NR1C1MGC2237, Nuclear receptor subfamily 1 group C member 1, peroxisome proliferator-activated receptor alpha, PPARalpha, PPAR-alpha, PPARMGC2452
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 145-420 of human PPAR alpha/NR1C1 (NP_005027) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism, mTOR Pathway, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 5465
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.